Assay Details
| Gene Symbol: LOC719797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847708.1 | XP_028703541.1 | 1630 | 151 | ||
| XM_028847709.1 | XP_028703542.1 | 3073 | 151 | |||
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194627.2 | NP_001181556.2 | 450 | 151 | ||
| XM_015135944.2 | XP_014991430.1 | 2028 | 151 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028847709.1 transcripts.
Set Membership: |
|

LOC719797
Genomic Map