Assay Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261705.1 | NP_001248634.1 | 182 | 113 | ||
XM_015114496.2 | XP_014969982.1 | 227 | 113 | |||
GenBank mRNA | JU332208.1 | - | 174 | 113 | ||
JU470462.1 | - | 157 | 113 | |||
JU470463.1 | - | 171 | 113 | |||
JV044622.1 | - | 182 | 113 | |||
JV567551.1 | - | 177 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261705.1, XM_015114496.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
