Assay Details
| Gene Symbol: CA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107970.4 | XP_001107970.2 | 236 | 81 | ||
| GenBank mRNA | JV628044.1 | - | 205 | 81 | ||
| JV695683.1 | - | 195 | 81 | |||
More Information
Set Membership: |
|

CA4
Genomic Map