Assay Details
| Gene Symbol: GABRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261668.1 | NP_001248597.1 | 615 | 84 | ||
| XM_015138315.2 | XP_014993801.1 | 758 | 84 | |||
| XM_015138316.2 | XP_014993802.1 | 826 | 84 | |||
| XM_028848079.1 | XP_028703912.1 | 536 | 84 | |||
| XM_028848080.1 | XP_028703913.1 | 825 | 84 | |||
| XM_028848081.1 | XP_028703914.1 | 1038 | 84 | |||
| GenBank mRNA | JU333815.1 | - | 591 | 84 | ||
| JU333816.1 | - | 556 | 84 | |||
| JU333817.1 | - | 615 | 84 | |||
| JV711309.1 | - | 565 | 84 | |||
More Information
Set Membership: |
|

GABRA2
Genomic Map