Assay Details
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113758.4 | XP_001113758.1 | 566 | 124 | ||
XM_015142222.2 | XP_014997708.1 | 468 | 124 | |||
XM_015142223.2 | XP_014997709.1 | 554 | 124 | |||
XM_015142224.2 | XP_014997710.1 | 392 | 124 | |||
XM_015142225.2 | XP_014997711.1 | 566 | 124 | |||
XM_015142226.2 | XP_014997712.1 | 566 | 124 | |||
XM_015142227.2 | XP_014997713.1 | 566 | 124 | |||
XM_028850902.1 | XP_028706735.1 | 468 | 124 | |||
GenBank mRNA | JU322759.1 | - | 441 | 124 | ||
JU322910.1 | - | 363 | 124 | |||
JU471948.1 | - | 371 | 124 | |||
JV567830.1 | - | 325 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
