Assay Details
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194014.2 | NP_001180943.1 | 143 | 107 | ||
XM_028835558.1 | XP_028691391.1 | 160 | 107 | |||
GenBank mRNA | JU321794.1 | - | 140 | 107 | ||
JU473739.1 | - | 136 | 107 | |||
JV045410.1 | - | 127 | 107 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
