Assay Details
Gene Symbol: DAZA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845077.1 | XP_028700910.1 | 1535 | 146 | ||
XM_028845135.1 | XP_028700968.1 | 1535 | 146 | |||
XM_028845166.1 | XP_028700999.1 | 1535 | 146 | |||
GenBank mRNA | FJ648738.1 | - | 928 | 146 |
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843651.1 | XP_028699484.1 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
