Assay Details
Gene Symbol: TAFA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130436.2 | XP_014985922.1 | 818 | 114 | ||
XM_028844354.1 | XP_028700187.1 | 431 | 114 | |||
XM_028844355.1 | XP_028700188.1 | 513 | 114 | |||
XM_028844356.1 | XP_028700189.1 | 295 | 114 | |||
XM_028844357.1 | XP_028700190.1 | 196 | 114 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015130436.2, XM_028844354.1, XM_028844355.1, XM_028844356.1, XM_028844357.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
