Assay Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146365.2 | XP_015001851.1 | 376 | 135 | ||
GenBank mRNA | JU332950.1 | - | 331 | 135 | ||
JU474334.1 | - | 351 | 135 | |||
JV046209.1 | - | 360 | 135 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
