Assay Details
| Gene Symbol: LOC709083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146586.2 | XP_015002072.1 | 2003 | 119 | ||
| XM_015146587.2 | XP_015002073.1 | 1977 | 119 | |||
| XM_015146590.2 | XP_015002076.1 | 2003 | 119 | |||
| XM_015146591.2 | XP_015002077.1 | 1575 | 119 | |||
| XM_015146593.2 | XP_015002079.1 | 1907 | 119 | |||
| XM_015146594.2 | XP_015002080.1 | 2003 | 119 | |||
| XM_015146595.2 | XP_015002081.1 | 1400 | 119 | |||
| XM_028826047.1 | XP_028681880.1 | 1777 | 119 | |||
| XM_028826048.1 | XP_028681881.1 | 2104 | 119 | |||
| XM_028826050.1 | XP_028681883.1 | 2003 | 119 | |||
| XM_028826051.1 | XP_028681884.1 | 2207 | 119 | |||
| XM_028826052.1 | XP_028681885.1 | 1582 | 119 | |||
| XM_028826053.1 | XP_028681886.1 | 2123 | 119 | |||
| XM_028826054.1 | XP_028681887.1 | 2449 | 119 | |||
| GenBank mRNA | JU348877.1 | - | 1661 | 119 | ||
| JU477307.1 | - | 1617 | 119 | |||
More Information
Set Membership: |
|

LOC709083
Genomic Map