Assay Details
| Gene Symbol: SGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089281.4 | XP_001089281.2 | 1646 | 93 | ||
| XM_028843647.1 | XP_028699480.1 | 848 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SGO1| Gene Symbol: SGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089281.4 | XP_001089281.2 | 1646 | 93 | ||
| XM_028843647.1 | XP_028699480.1 | 848 | 93 | |||
Set Membership: |
|