Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137300.2 | XP_014992786.1 | 768 | 123 | ||
| GenBank mRNA | JV047015.1 | - | 261 | 123 | ||
| JV636144.1 | - | 395 | 123 | |||
More Information
Set Membership: |
|

CENPW
Genomic Map