Assay Details
Gene Symbol: SLC3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113953.2 | XP_014969439.2 | 1172 | 78 | ||
XM_028833333.1 | XP_028689166.1 | 1024 | 78 | |||
XM_028833334.1 | XP_028689167.1 | 968 | 78 | |||
GenBank mRNA | JU323244.1 | - | 720 | 78 | ||
JU323362.1 | - | 1135 | 78 | |||
JU471247.1 | - | 1054 | 78 | |||
JV043763.1 | - | 1014 | 78 | |||
JV527768.1 | - | 1046 | 78 | |||
JV626278.1 | - | 654 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
