Assay Details
| Gene Symbol: GDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094714.4 | XP_001094714.2 | 1709 | 102 | ||
| XM_015139905.2 | XP_014995391.1 | 833 | 102 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDNF| Gene Symbol: GDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094714.4 | XP_001094714.2 | 1709 | 102 | ||
| XM_015139905.2 | XP_014995391.1 | 833 | 102 | |||
Set Membership: |
|