Assay Details
| Gene Symbol: TMIGD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194531.2 | NP_001181460.1 | 852 | 87 | ||
| XM_015118862.2 | XP_014974348.2 | 764 | 87 | |||
| XM_015118863.2 | XP_014974349.1 | 760 | 87 | |||
More Information
Set Membership: |
|

TMIGD1
Genomic Map