Assay Details
| Gene Symbol: C1H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090172.4 | XP_001090172.1 | 558 | 115 | ||
| XM_028849240.1 | XP_028705073.1 | 431 | 115 | |||
More Information
Set Membership: |
|

C1H1orf100
Genomic Map