Assay Details
| Gene Symbol: LOC709576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107762.4 | XP_001107762.1 | 576 | 72 | ||
| XM_001107827.4 | XP_001107827.1 | 585 | 72 | |||
| XM_001107893.4 | XP_001107893.1 | 742 | 72 | |||
| XM_015126516.2 | XP_014982002.1 | 653 | 72 | |||
| XM_028840706.1 | XP_028696539.1 | 688 | 72 | |||
| XM_028840707.1 | XP_028696540.1 | 602 | 72 | |||
| XM_028840708.1 | XP_028696541.1 | 694 | 72 | |||
| XM_028840709.1 | XP_028696542.1 | 776 | 72 | |||
| XM_028840710.1 | XP_028696543.1 | 707 | 72 | |||
| GenBank mRNA | JU321341.1 | - | 559 | 72 | ||
| JU472551.1 | - | 556 | 72 | |||
| JV044486.1 | - | 561 | 72 | |||
| JV570069.1 | - | 133 | 72 | |||

LOC709576
Genomic Map