Assay Details
Gene Symbol: TAF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140184.2 | XP_014995670.1 | 371 | 111 | ||
GenBank mRNA | JU476968.1 | - | 353 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015140184.2 transcripts.
Set Membership: |
![]() ![]() |
