Assay Details
Gene Symbol: LRIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805650.2 | XP_002805696.2 | 110 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
