Assay Details
| Gene Symbol: POLR2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142334.2 | XP_014997820.1 | 1280 | 123 | ||
| GenBank mRNA | JU336532.1 | - | 1084 | 123 | ||
| JU477344.1 | - | 1095 | 123 | |||
| JV048334.1 | - | 1083 | 123 | |||
| JV724274.1 | - | 1105 | 123 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POLR2M
Genomic Map