Assay Details
| Gene Symbol: LOC712294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141725.2 | XP_014997211.2 | 4358 | 141 | ||
| XM_015141726.2 | XP_014997212.2 | 4343 | 141 | |||
| XM_015141727.2 | XP_014997213.2 | 2435 | 141 | |||
| XM_015141728.2 | XP_014997214.2 | 1822 | 141 | |||
| XM_028850610.1 | XP_028706443.1 | 4119 | 141 | |||
| XM_028850611.1 | XP_028706444.1 | 3803 | 141 | |||
| GenBank mRNA | JU321636.1 | - | 4318 | 141 | ||
| JU336956.1 | - | 4303 | 141 | |||
| JV726178.1 | - | 4318 | 141 | |||
More Information
Set Membership: |
|

LOC712294
Genomic Map