Assay Details
Gene Symbol: LOC712294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141725.2 | XP_014997211.2 | 4624 | 92 | ||
XM_015141726.2 | XP_014997212.2 | 4609 | 92 | |||
XM_015141727.2 | XP_014997213.2 | 2701 | 92 | |||
XM_015141728.2 | XP_014997214.2 | 2088 | 92 | |||
XM_028850610.1 | XP_028706443.1 | 4385 | 92 | |||
XM_028850611.1 | XP_028706444.1 | 4069 | 92 | |||
GenBank mRNA | JU321636.1 | - | 4584 | 92 | ||
JU336956.1 | - | 4569 | 92 | |||
JV726178.1 | - | 4584 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
