Assay Details
| Gene Symbol: COX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257834.1 | NP_001244763.1 | 282 | 155 | ||
| GenBank mRNA | JU323489.1 | - | 282 | 155 | ||
| JU477741.1 | - | 219 | 155 | |||
| JV048636.1 | - | 212 | 155 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COX19
Genomic Map