Assay Details
Gene Symbol: CRELD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266467.1 | NP_001253396.1 | 799 | 71 | ||
XM_015130259.2 | XP_014985745.1 | 435 | 71 | |||
XM_015130260.2 | XP_014985746.1 | 340 | 71 | |||
XM_015130261.2 | XP_014985747.1 | 411 | 71 | |||
GenBank mRNA | JU336330.1 | - | 700 | 71 | ||
JU477153.1 | - | 799 | 71 | |||
JV045709.1 | - | 799 | 71 | |||
JV600738.1 | - | 797 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
