Assay Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112536.2 | XP_014968022.1 | 300 | 109 | ||
XM_028831531.1 | XP_028687364.1 | 268 | 109 | |||
GenBank mRNA | JV565023.1 | - | 197 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
