Assay Details
Gene Symbol: PXYLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171842.1 | NP_001165313.1 | 651 | 134 | ||
XM_015131963.2 | XP_014987449.1 | 388 | 134 | |||
XM_028843385.1 | XP_028699218.1 | 311 | 134 | |||
XM_028843386.1 | XP_028699219.1 | 270 | 134 | |||
XM_028843387.1 | XP_028699220.1 | 646 | 134 | |||
GenBank mRNA | AF218042.1 | - | 651 | 134 | ||
JU326672.1 | - | 198 | 134 | |||
JU477839.1 | - | 312 | 134 | |||
JV633483.1 | - | 357 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001171842.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
