Assay Details
Gene Symbol: MAJIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113829.2 | XP_014969315.2 | 838 | 122 | ||
XM_015113830.2 | XP_014969316.2 | 712 | 122 | |||
XM_015113832.2 | XP_014969318.1 | 774 | 122 | |||
XM_028833250.1 | XP_028689083.1 | 664 | 122 | |||
XM_028833251.1 | XP_028689084.1 | 733 | 122 | |||
XM_028833252.1 | XP_028689085.1 | 701 | 122 | |||
XM_028833253.1 | XP_028689086.1 | 534 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
