Assay Details
| Gene Symbol: PHYH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195489.1 | NP_001182418.1 | 246 | 72 | ||
| GenBank mRNA | HM355984.1 | - | 246 | 72 | ||
| JU323344.1 | - | 259 | 72 | |||
| JV043312.1 | - | 252 | 72 | |||
More Information
Set Membership: |
|

PHYH
Genomic Map