Assay Details
| Gene Symbol: IL18BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123353.1 | NP_001116825.1 | 352 | 117 | ||
| XM_015115021.2 | XP_014970507.1 | 1049 | 117 | |||
| XM_015115022.1 | XP_014970508.1 | 1489 | 117 | |||
| XM_015115023.1 | XP_014970509.1 | 1601 | 117 | |||
| GenBank mRNA | EU564533.1 | - | 352 | 117 | ||
| JU478487.1 | - | 524 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL18BP
Genomic Map