Assay Details
Gene Symbol: CCNI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140763.2 | XP_014996249.2 | 1675 | 84 | ||
GenBank mRNA | JU327878.1 | - | 1422 | 84 | ||
JU327879.1 | - | 1423 | 84 |
Gene Symbol: SEPTIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV710337.1 | - | 2838 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
