Assay Details
| Gene Symbol: ARHGAP11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084475.4 | XP_001084475.3 | 1745 | 120 | ||
| XM_028850693.1 | XP_028706526.1 | 1746 | 120 | |||
| XM_028850694.1 | XP_028706527.1 | 1750 | 120 | |||
| GenBank mRNA | JU323442.1 | - | 1306 | 120 | ||
| JU473039.1 | - | 1591 | 120 | |||
More Information
Set Membership: |
|

ARHGAP11A
Genomic Map