Assay Details
Gene Symbol: CD37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114865.4 | XP_001114865.2 | 629 | 106 | ||
XM_015124563.2 | XP_014980049.1 | 583 | 106 | |||
XM_028839715.1 | XP_028695548.1 | 631 | 106 | |||
GenBank mRNA | JU323266.1 | - | 562 | 106 | ||
JU470714.1 | - | 573 | 106 | |||
JU496668.1 | - | 569 | 106 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
