Assay Details
| Gene Symbol: CKLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126320.2 | XP_014981806.1 | 257 | 133 | ||
| XM_015126321.2 | XP_014981807.1 | 233 | 133 | |||
| GenBank mRNA | JU471341.1 | - | 217 | 133 | ||
| JU471342.1 | - | 217 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CKLF
Genomic Map