Assay Details
| Gene Symbol: AOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100309.4 | XP_001100309.1 | 1970 | 59 | ||
| XM_015135278.2 | XP_014990764.1 | 1942 | 59 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AOC1| Gene Symbol: AOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100309.4 | XP_001100309.1 | 1970 | 59 | ||
| XM_015135278.2 | XP_014990764.1 | 1942 | 59 | |||
Set Membership: |
|