Assay Details
Gene Symbol: CST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113660.2 | XP_014969146.1 | 930 | 116 | ||
GenBank mRNA | JV044001.1 | - | 404 | 116 | ||
JV635664.1 | - | 389 | 116 | |||
JV707621.1 | - | 351 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
