Assay Details
Gene Symbol: LOC114675527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842746.1 | XP_028698579.1 | 478 | 89 |
Gene Symbol: ACP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001357040.1 | NP_001343969.1 | 528 | 89 | ||
NM_001357041.1 | NP_001343970.1 | 528 | 89 | |||
GenBank mRNA | JU326844.1 | - | 446 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
