Assay Details
| Gene Symbol: APEX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258109.1 | NP_001245038.1 | 266 | 127 | ||
| XM_015143085.2 | XP_014998571.1 | 512 | 127 | |||
| XM_015143087.2 | XP_014998573.1 | 530 | 127 | |||
| XM_015143088.2 | XP_014998574.1 | 516 | 127 | |||
| XM_015143089.2 | XP_014998575.1 | 506 | 127 | |||
| XM_015143090.2 | XP_014998576.1 | 400 | 127 | |||
| XM_015143091.2 | XP_014998577.1 | 404 | 127 | |||
| GenBank mRNA | JU326201.1 | - | 182 | 127 | ||
| JU475401.1 | - | 266 | 127 | |||
| JV046924.1 | - | 502 | 127 | |||
| JV046925.1 | - | 436 | 127 | |||
| JV046926.1 | - | 311 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

APEX1
Genomic Map