Assay Details
| Gene Symbol: CCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265987.1 | NP_001252916.1 | 851 | 104 | ||
| XM_028846630.1 | XP_028702463.1 | 1010 | 104 | |||
| XM_028846631.1 | XP_028702464.1 | 794 | 104 | |||
| GenBank mRNA | JU322013.1 | - | 851 | 104 | ||
| JU478174.1 | - | 608 | 104 | |||
| JV048616.1 | - | 851 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCND3
Genomic Map