Assay Details
| Gene Symbol: CDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082641.4 | XP_001082641.1 | 1590 | 63 | ||
| GenBank mRNA | JV639598.1 | - | 1498 | 63 | ||
| JV704794.1 | - | 1498 | 63 | |||
More Information
Set Membership: |
|

CDH5
Genomic Map