Assay Details
| Gene Symbol: LOC722291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116461.2 | XP_014971947.2 | 1828 | 87 | ||
| XM_015116463.2 | XP_014971949.2 | 1585 | 87 | |||
| GenBank mRNA | JV640344.1 | - | 150 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015116463.2 transcripts.
Set Membership: |
|

LOC722291
Genomic Map