Assay Details
| Gene Symbol: LOC722291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV640344.1 | - | 714 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC722291| Gene Symbol: LOC722291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV640344.1 | - | 714 | 108 | ||
Set Membership: |
|