Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099283.4 | XP_001099283.3 | 2430 | 105 | ||
| XM_015138618.2 | XP_014994104.2 | 2430 | 105 | |||
| XM_015138619.2 | XP_014994105.2 | 2300 | 105 | |||
| GenBank mRNA | JU332547.1 | - | 2366 | 105 | ||
| JU474019.1 | - | 2373 | 105 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CENPC
Genomic Map