Assay Details
| Gene Symbol: LOC114675313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842554.1 | XP_028698387.1 | 483 | 142 | ||
| Gene Symbol: CNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261809.1 | NP_001248738.1 | 730 | 142 | ||
| XM_015145070.2 | XP_015000556.1 | 569 | 142 | |||
| GenBank mRNA | JU319626.1 | - | 633 | 142 | ||
| JU472900.1 | - | 673 | 142 | |||
| JV044008.1 | - | 730 | 142 | |||
| JV451657.1 | - | 748 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC114675313
Genomic Map