Assay Details
| Gene Symbol: COX4I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU474337.1 | - | 119 | 143 | ||
| JV046213.1 | - | 143 | ||||
| JV046215.1 | - | 143 | ||||
| JV673172.1 | - | 143 | ||||
More Information
Set Membership: |
|

COX4I1
Genomic Map