Assay Details
| Gene Symbol: FRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112190.4 | XP_001112190.1 | 1706 | 89 | ||
| XM_015137546.2 | XP_014993032.1 | 1420 | 89 | |||
| XM_015137548.2 | XP_014993034.1 | 777 | 89 | |||
| XM_015137549.2 | XP_014993035.1 | 723 | 89 | |||
| XM_028847052.1 | XP_028702885.1 | 1709 | 89 | |||
| XM_028847053.1 | XP_028702886.1 | 1917 | 89 | |||
More Information
Set Membership: |
|

FRK
Genomic Map