Assay Details
Gene Symbol: KLRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032828.1 | NP_001028000.1 | 360 | 100 | ||
XM_015150987.2 | XP_015006473.1 | 618 | 100 | |||
XM_015150989.2 | XP_015006475.1 | 369 | 100 | |||
GenBank mRNA | AF190931.1 | - | 179 | 100 | ||
AF190932.1 | - | 179 | 100 | |||
AF190933.1 | - | 86 | 100 | |||
AF294886.1 | - | 360 | 100 | |||
AJ585527.1 | - | 200 | 100 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
