Assay Details
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114669.2 | XP_014970155.2 | 407 | 113 | ||
XM_028833571.1 | XP_028689404.1 | 392 | 113 | |||
XM_028833572.1 | XP_028689405.1 | 274 | 113 | |||
XM_028833573.1 | XP_028689406.1 | 386 | 113 | |||
XM_028833574.1 | XP_028689407.1 | 392 | 113 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
