Assay Details
Gene Symbol: LOC696037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266719.2 | NP_001253648.1 | 100 | 123 | ||
XM_015146113.2 | XP_015001599.1 | 699 | 123 | |||
GenBank mRNA | JU322979.1 | - | 203 | 123 | ||
JU471851.1 | - | 101 | 123 | |||
JV616749.1 | - | 207 | 123 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001266719.2, XM_015146113.2 transcripts.
This assay is in the 5'UTR region of the NM_001266719.2, XM_015146113.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
