Assay Details
| Gene Symbol: LY6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266030.1 | NP_001252959.1 | 251 | 79 | ||
| XM_015146115.2 | XP_015001601.1 | 231 | 79 | |||
| XM_028852248.1 | XP_028708081.1 | 413 | 79 | |||
| XM_028852249.1 | XP_028708082.1 | 360 | 79 | |||
| GenBank mRNA | JU322432.1 | - | 251 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LY6H
Genomic Map