Assay Details
| Gene Symbol: NDUFS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266182.1 | NP_001253111.1 | 256 | 141 | ||
| XM_015113462.2 | XP_014968948.1 | 411 | 141 | |||
| XM_015113463.1 | XP_014968949.1 | 338 | 141 | |||
| GenBank mRNA | JU323320.1 | - | 261 | 141 | ||
| JU475708.1 | - | 261 | 141 | |||
| JV044651.1 | - | 256 | 141 | |||
| JV617627.1 | - | 284 | 141 | |||
More Information
Set Membership: |
|

NDUFS8
Genomic Map